Merck
  • Cationic palladium(II) catalysis: C-H activation/Suzuki-Miyaura couplings at room temperature.
Cationic palladium(II) catalysis: C-H activation/Suzuki-Miyaura couplings at room temperature.
Journal of the American Chemical Society (2010-03-24)
Takashi Nishikata, Alexander R Abela, Shenlin Huang, Bruce H Lipshutz
ABSTRACT
Cationic palladium(II) catalyst realized facile C-H activation of aryl urea with arylboronic acids at room temperature. This reaction is extremely mild to carry out aromatic C-H activations through electrophilic substitution.
Sigma-Aldrich
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate