Merck
  • Rho-associated protein kinase 2 (ROCK2): a new target of autoimmunity in paraneoplastic encephalitis.
Rho-associated protein kinase 2 (ROCK2): a new target of autoimmunity in paraneoplastic encephalitis.
Acta neuropathologica communications (2017-05-31)
Stoyan Popkirov, Ilya Ayzenberg, Stefanie Hahn, Jan Bauer, Yvonne Denno, Nicole Rieckhoff, Christiane Radzimski, Volkmar H Hans, Sebastian Berg, Florian Roghmann, Joachim Noldus, Christian G Bien, Sabine Skodda, Jörg Wellmer, Winfried Stöcker, Christos Krogias, Ralf Gold, Uwe Schlegel, Christian Probst, Lars Komorowski, Ramona Miske, Ingo Kleiter
ABSTRACT
Onconeural antibodies are associated with cancer and paraneoplastic encephalitis. While their pathogenic role is still largely unknown, their high diagnostic value is undisputed. In this study we describe the discovery of a novel target of autoimmunity in an index case of paraneoplastic encephalitis associated with urogenital cancer.A 75-year-old man with a history of invasive bladder carcinoma 6 years ago with multiple recurrences and a newly discovered renal cell carcinoma presented with seizures and progressive cognitive decline followed by super-refractory status epilepticus. Clinical and ancillary findings including brain biopsy suggested paraneoplastic encephalitis. Immunohistochemistry of the brain biopsy was used to characterize the inflammatory response. Indirect immunofluorescence assay (IFA) was used for autoantibody screening. The autoantigen was identified by histo-immunoprecipitation and mass spectrometry and was validated by expressing the recombinant antigen in HEK293 cells and neutralization tests. Sera from 125 control patients were screened using IFA to test for the novel autoantibodies.IFA analysis of serum revealed a novel autoantibody against brain tissue. An intracellular enzyme, Rho-associated protein kinase 2 (ROCK2), was identified as target-antigen. ROCK2 was expressed in affected brain tissue and archival bladder tumor samples of this patient. Brain histopathology revealed appositions of cytotoxic CD8
Sigma-Aldrich
Anti-ROCK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.