Merck
  • Hepatic thyroid signaling of heat-stressed late pregnant and early lactating cows.
Hepatic thyroid signaling of heat-stressed late pregnant and early lactating cows.
The Journal of endocrinology (2017-05-14)
Joachim M Weitzel, Torsten Viergutz, Dirk Albrecht, Rupert Bruckmaier, Marion Schmicke, Armin Tuchscherer, Franziska Koch, Björn Kuhla
ABSTRACT
During the transition between late gestation and early lactation, dairy cows experience severe metabolic stress due to the high energy and nutrient requirements of the fetus and the mammary gland. Additional thermal stress that occurs with rising temperatures during the ongoing climate change has further adverse implications on energy intake, metabolism and welfare. The thyroid hormone (TH)-mediated cellular signaling has a pivotal role in regulation of body temperature, energy intake and metabolic adaptation to heat. To distinguish between energy intake and heat stress-related effects, Holstein cows were first kept at thermoneutrality at 15°C followed by exposure to heat stress (HS) at 28°C or pair-feeding (PF) at 15°C for 6 days, in late pregnancy and again in early lactation. Herein, we focused on hepatic metabolic changes associated with alterations in the hypothalamic-pituitary-thyroid axis in HS and PF animals. T
Sigma-Aldrich
Anti-P4HB (AB2) antibody produced in rabbit, affinity isolated antibody