- Insights into substrate binding of ferulic acid esterases by arabinose and methyl hydroxycinnamate esters and molecular docking.
Insights into substrate binding of ferulic acid esterases by arabinose and methyl hydroxycinnamate esters and molecular docking.
Ferulic acid esterases (FAE, EC 3.1.1.73) cleave the arabinose hydroxycinnamate ester in plant hemicellulose and other related substrates. FAE are commonly categorised as type A-D based on catalytic activities towards model, short alkyl chain esters of hydroxycinnamates. However, this system correlates poorly with sequence and structural features of the enzymes. In this study, we investigated the basis of the type A categorisation of an FAE from Aspergillus niger, AnFaeA, by comparing its activity toward methyl and arabinose hydroxycinnamate esters. k