- APRDX1 mutant allele causes a MMACHC secondary epimutation in cblC patients.
APRDX1 mutant allele causes a MMACHC secondary epimutation in cblC patients.
Nature communications (2018-01-06)
Jean-Louis Guéant, Céline Chéry, Abderrahim Oussalah, Javad Nadaf, David Coelho, Thomas Josse, Justine Flayac, Aurélie Robert, Isabelle Koscinski, Isabelle Gastin, Pierre Filhine-Tresarrieu, Mihaela Pupavac, Alison Brebner, David Watkins, Tomi Pastinen, Alexandre Montpetit, Fadi Hariri, David Tregouët, Benjamin A Raby, Wendy K Chung, Pierre-Emmanuel Morange, D Sean Froese, Matthias R Baumgartner, Jean-François Benoist, Can Ficicioglu, Virginie Marchand, Yuri Motorin, Chrystèle Bonnemains, François Feillet, Jacek Majewski, David S Rosenblatt
PMID29302025
ABSTRACT
To date, epimutations reported in man have been somatic and erased in germlines. Here, we identify a cause of the autosomal recessive cblC class of inborn errors of vitamin B
MATERIALS