Merck
NiCo
Nanoscale (2017-11-28)
Balasubramaniam Saravanakumar, Santhana Sivabalan Jayaseelan, Min-Kang Seo, Hak-Yong Kim, Byoung-Suhk Kim
ABSTRACT
Wire type supercapacitors with high energy and power densities have generated considerable interest in wearable applications. Herein, we report a novel NiCo
Sigma-Aldrich
Cyanamide solution, 50 wt. % in H2O
Pricing and availability is not currently available.