Merck
  • Function and structure relationships of a β-1,2-glucooligosaccharide-degrading β-glucosidase.
Function and structure relationships of a β-1,2-glucooligosaccharide-degrading β-glucosidase.
FEBS letters (2017-11-14)
Rikuto Ishiguro, Nobukiyo Tanaka, Koichi Abe, Masahiro Nakajima, Takuma Maeda, Akimasa Miyanaga, Yuta Takahashi, Naohisa Sugimoto, Hiroyuki Nakai, Hayao Taguchi
ABSTRACT
BT_3567 protein, a putative β-glucosidase from Bacteroides thetaiotaomicron, exhibits higher activity toward Sop
Sigma-Aldrich
β-Gentiobiose, ≥85% (remainder primarily α-anomer)