Merck
  • High-Resolution, Accurate-Mass (HRAM) Mass Spectrometry Urine Steroid Profiling in the Diagnosis of Adrenal Disorders.
High-Resolution, Accurate-Mass (HRAM) Mass Spectrometry Urine Steroid Profiling in the Diagnosis of Adrenal Disorders.
Clinical chemistry (2017-08-18)
Jolaine M Hines, Irina Bancos, Cristian Bancos, Raman D Singh, Aditya V Avula, William F Young, Stefan K Grebe, Ravinder J Singh
ABSTRACT
Steroid profiling is a promising diagnostic tool with adrenal tumors, Cushing syndrome (CS), and disorders of steroidogenesis. Our objective was to develop a multiple-steroid assay using liquid-chromatography, high-resolution, accurate-mass mass spectrometry (HRAM LC-MS) and to validate the assay in patients with various adrenal disorders. We collected 24-h urine samples from 114 controls and 71 patients with adrenal diseases. An HRAM LC-MS method was validated for quantitative analysis of 26 steroid metabolites in hydrolyzed urine samples. Differences in steroid excretion between patients were analyzed based on Limits of quantification were 20 ng/mL. Dilution linearity ranged from 80% to 120% with means of 93% to 110% for all but 2 analytes. Intraassay and interassay imprecision ranged from 3% to 18% for all but 1 analyte. Control women had lower excretion of androgen and glucocorticoid precursors/metabolites than men ( Our HRAM LC-MS assay successfully quantifies 26 steroids in urine. The statistically significant differences in steroid production of ACC vs ACA, adrenal vs pituitary CS, and in congenital adrenal hyperplasia should allow for improved diagnosis of patients with these diseases.
Sigma-Aldrich
Dehydroepiandrosterone-2,2,3,4,4,6-d6, 97 atom % D, 98% (CP)
Supelco
Androsterone, VETRANAL®, analytical standard
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%