Merck
  • Automated microscopy screening for compounds that partially revert cholesterol accumulation in Niemann-Pick C cells.
Automated microscopy screening for compounds that partially revert cholesterol accumulation in Niemann-Pick C cells.
Journal of lipid research (2005-11-17)
Nina H Pipalia, Amy Huang, Harold Ralph, Madalina Rujoi, Frederick R Maxfield
ABSTRACT
Niemann-Pick disease type C (NPC) is an autosomal recessive genetic disorder manifested by abnormal accumulation of unesterified cholesterol and other lipids. We screened combinatorially synthesized chemical libraries to identify compounds that would partially revert cholesterol accumulation. Cultured CHO cells with NPC phenotypes (CT60 and CT43) were used for screening along with normal CHO cells as a control. We developed an automated microscopy assay based on imaging of filipin fluorescence for estimating cholesterol accumulation in lysosomal storage organelles. Our primary screen of 14,956 compounds identified 14 hit compounds that caused significant reduction in cellular cholesterol accumulation at 10 microM. We then screened a secondary library of 3,962 compounds selected based on chemical similarity to the initial hits and identified 7 compounds that demonstrated greater efficacy and lower toxicity than the original hits. These compounds are effective at concentrations of 123 nM to 3 microM in reducing the cholesterol accumulation in cells with a NPC1 phenotype.
Sigma-Aldrich
Filipin complex ready made solution, from Streptomyces filipinensis, 5mg/ml in DMSO based solution
Pricing and availability is not currently available.
Sigma-Aldrich
Filipin III ready made solution, from Streptomyces filipinensis, 1mg/ml in DMSO based solution
Pricing and availability is not currently available.