Merck
Involvement of dysregulated IK
Placenta (2017-10-01)
Fan-Fan Li, Meng-Zhou He, Yin Xie, Yuan-Yuan Wu, Mei-Tao Yang, Yao Fan, Fu-Yuan Qiao, Dong-Rui Deng
ABSTRACT
Excessive constriction of placental chorionic plate arteries (CPAs) may be associated with preeclampsia (PE). Nitric oxide (NO) as well as intermediate and small Ca The location of IK IK This study provides evidence that dysregulated IK
Sigma-Aldrich
NS309, ≥98% (HPLC), solid