- Adaptable interaction between aquaporin-1 and band 3 reveals a potential role of water channel in blood CO
Adaptable interaction between aquaporin-1 and band 3 reveals a potential role of water channel in blood CO
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-06-10)
Kate Hsu, Ting-Ying Lee, Ammasi Periasamy, Fu-Jen Kao, Li-Tzu Li, Chuang-Yu Lin, Hui-Ju Lin, Marie Lin
PMID28596233
ABSTRACT
Human CO
Sigma-Aldrich
4-(2-[6-(Dioctylamino)-2-naphthalenyl]ethenyl)-1-(3-sulfopropyl)pyridinium inner salt, ≥95% (HPLC), solid