- Effect of xylanase and α-amylase on DON and its conjugates during the breadmaking process.
Effect of xylanase and α-amylase on DON and its conjugates during the breadmaking process.
Deoxynivalenol (DON) is one of the most frequently occurring mycotoxins in wheat crops worldwide and poses a risk to human and animal health due to its wide range of adverse effects. Deoxynivalenol-3-glucoside (DON-3-glucoside) is a DON plant conjugate that is widely found in cereal products. As DON accumulation in the field seems unavoidable, it is important to investigate all of the conditions that affect its stability during food processing. One of the most consumed cereal product around the world is bread, however the published information about DON stability in bread shows a large variability of results because a huge amount of factors affect DON and its modified forms. So, the aim of this research was to study the fate of DON and its modified forms through the breadmaking process with the addition of xylanase and α-amylase at different fermentation temperatures. Moreover, different α-amylase and xylanase concentrations were added to the dough to be fermented. To quantify DON and its derived forms in the samples, liquid chromatography with double mass spectrophotometer was used. DON was reduced during fermentation and baking; however, the reduction at each step was related to the fermentation temperature. The presence of α-amylase and xylanase caused increases in DON during fermentation and during early baking. DON-3-glucoside was slightly reduced after fermentation and was widely increased (>80%) after baking. Deepoxy-deoxynivalenol (DOM-1) increased during the breadmaking process. Breadmaking process can reduce DON concentration, however xylanase and α-amylase presence cause increases of DON.