Merck
  • The single IGF-1 partial deficiency is responsible for mitochondrial dysfunction and is restored by IGF-1 replacement therapy.
The single IGF-1 partial deficiency is responsible for mitochondrial dysfunction and is restored by IGF-1 replacement therapy.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society (2017-06-27)
M Olleros Santos-Ruiz, M C Sádaba, I Martín-Estal, U Muñoz, C Sebal Neira, I Castilla-Cortázar
ABSTRACT
We previously described in cirrhosis and aging, both conditions of IGF-1 deficiency, a clear hepatic mitochondrial dysfunction with increased oxidative damage. In both conditions, the hepatic mitochondrial function was improved with low doses of IGF-1. The aim of this work was to explore if the only mere IGF-1 partial deficiency, without any exogenous insult, is responsible for hepatic mitochondrial dysfunction. Heterozygous (igf1 Hz mice showed a significant reduction in hepatic mitochondrial membrane potential (MMP) and ATPase activity, and an increase in intramitochondrial free radical production and proton leak rates, compared to controls. These parameters were normalized by IGF-1 replacement therapy. No significant differences were found between groups in oxygen consumption and antioxidant enzyme activities, except for catalase, whose activity was increased in both Hz groups. Relevant genes coding for proteins involved in mitochondrial protection and survival were altered in Hz group and were reverted to normal in Hz+IGF-1 group. The mere IGF-1 partial deficiency is per se associated with hepatic mitochondrial dysfunction sensitive to IGF-1 replacement therapy. Results in this work prove that IGF-1 is involved in hepatic mitochondrial protection, because it is able to reduce free radical production, oxidative damage and apoptosis. All these IGF-1 actions are mediated by the modulation of the expression of genes encoding citoprotective and antiapoptotic proteins.
Sigma-Aldrich
1-Methyl-2-phenylindole, 99%