Merck
  • B4G2 induces mitochondrial apoptosis by the ROS-mediated opening of Ca(2+)-dependent permeability transition pores.
B4G2 induces mitochondrial apoptosis by the ROS-mediated opening of Ca(2+)-dependent permeability transition pores.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-09-19)
Nan Yao, Ying-Jie Li, Dong-Mei Zhang, Dao-Lu Liu, Ming-Kuen Tang, Anita Yiu, Yong Li, Wei-Min Chen, Ping Lan, Zhe Yao, Zhe-Sheng Chen, Wen-Cai Ye
ABSTRACT
Hepatocellular carcinoma (HCC) is the most common type of liver cancer. At present, only sorafenib is approved to treat HCC. In this study, we found that a 23-hydroxybetulinic acid derivative, B4G2, exhibited potent antiproliferative activity in HCC cell lines. We used four HCC cell lines (HepG2, HepG2/ADM, Hep3B and Bel-7402) to evaluate the anti-tumour activity and explore underlying mechanisms by which B4G2 induces apoptosis. Among these cell lines, HepG2 showed the highest sensitivity to B4G2. HepG2 cells treated with B4G2 showed a depolarized mitochondrial membrane potential, released cytochrome c, activated caspase-9 and caspase-3 and cleaved poly ADP-ribose polymerase (PARP). However, Z-VAD-FMK, a pan-caspase inhibitor, did not attenuate B4G2-induced apoptosis, implying that the induction of mitochondrial apoptosis by B4G2 may be independent of caspases. Moreover, pre-treatment with MgCl2, a blocker of Ca2+-dependent permeability transition (PT) pores, attenuated the depolarization of the mitochondrial potential and decreased the population of apoptotic cells, indicating that B4G2-induced apoptosis was partly dependent on the opening of the Ca2+-dependent PT pores. B4G2 also increased the levels of intracellular calcium and reactive oxygen species (ROS). Furthermore, an ROS scavenger, N-acetyl-cysteine (NAC), markedly decreased the accumulation of intracellular calcium and apoptosis. This is the first demonstration that B4G2 inhibits the growth of HCC cells and induces mitochondrial apoptosis in hepatocellular carcinoma cells by the ROS-mediated opening of Ca2+-dependent permeability transition pores.
Sigma-Aldrich
23-Hydroxybetulinic acid, ≥98% (HPLC)
Pricing and availability is not currently available.