Merck
Europium-doped NaGd(WO
Dalton transactions (Cambridge, England : 2003) (2017-08-08)
Mariano Laguna, Alberto Escudero, Nuria O Núñez, Ana I Becerro, Manuel Ocaña
ABSTRACT
Uniform Eu-doped NaGd(WO
Sigma-Aldrich
Europium(III) nitrate pentahydrate, 99.9% trace metals basis
Pricing and availability is not currently available.