Merck
  • Comparing DNA enrichment of proliferating cells following administration of different stable isotopes of heavy water.
Comparing DNA enrichment of proliferating cells following administration of different stable isotopes of heavy water.
Scientific reports (2017-06-24)
Don E Farthing, Nataliya P Buxbaum, Philip J Lucas, Natella Maglakelidze, Brittany Oliver, Jiun Wang, Kevin Hu, Ehydel Castro, Catherine V Bare, Ronald E Gress
ABSTRACT
Deuterated water (
Sigma-Aldrich
2′-Deoxyadenosine monohydrate, ≥99%