Merck
  • Luman contributes to brefeldin A-induced prion protein gene expression by interacting with the ERSE26 element.
Luman contributes to brefeldin A-induced prion protein gene expression by interacting with the ERSE26 element.
Scientific reports (2017-02-17)
Marc-André Déry, Andréa C LeBlanc
ABSTRACT
The cellular prion protein (PrP) is essential for transmissible prion diseases, but its exact physiological function remains unclear. Better understanding the regulation of the human prion protein gene (PRNP) expression can provide insight into this elusive function. Spliced XBP1 (sXBP1) was recently shown to mediate endoplasmic reticulum (ER) stress-induced PRNP expression. In this manuscript, we identify Luman, a ubiquitous, non-canonical unfolded protein response (UPR), as a novel regulator of ER stress-induced PRNP expression. Luman activity was transcriptionally and proteolytically activated by the ER stressing drug brefeldin A (BFA) in human neurons, astrocytes, and breast cancer MCF-7 cells. Over-expression of active cleaved Luman (ΔLuman) increased PrP levels, while siRNA-mediated Luman silencing decreased BFA-induced PRNP expression. Site-directed mutagenesis and chromatin immunoprecipitation demonstrated that ΔLuman regulates PRNP expression by interacting with the ER stress response element 26 (ERSE26). Co-over-expression and siRNA-mediated silencing experiments showed that sXBP1 and ΔLuman both up-regulate ER stress-induced PRNP expression. Attempts to understand the function of PRNP up-regulation by Luman excluded a role in atorvastatin-induced neuritogenesis, ER-associated degradation, or proteasomal inhibition-induced cell death. Overall, these results refine our understanding of ER stress-induced PRNP expression and function.