Merck
  • Peripheral endocannabinoid signaling controls hyperphagia in western diet-induced obesity.
Peripheral endocannabinoid signaling controls hyperphagia in western diet-induced obesity.
Physiology & behavior (2017-01-10)
Donovan A Argueta, Nicholas V DiPatrizio
ABSTRACT
The endocannabinoid system in the brain and periphery plays a major role in controlling food intake and energy balance. We reported that tasting dietary fats was met with increased levels of the endocannabinoids, 2-arachidonoyl-sn-glycerol (2-AG) and anandamide, in the rat upper small intestine, and pharmacological inhibition of this local signaling event dose-dependently blocked sham feeding of fats. We now investigated the contribution of peripheral endocannabinoid signaling in hyperphagia associated with chronic consumption of a western-style diet in mice ([WD] i.e., high fat and sucrose). Feeding patterns were assessed in male C57BL/6Tac mice maintained for 60days on WD or a standard rodent chow (SD), and the role for peripheral endocannabinoid signaling at CB
Sigma-Aldrich
AM6545, ≥98% (HPLC)
Pricing and availability is not currently available.