Merck
  • Epitranscriptomic Enhancement of Influenza A Virus Gene Expression and Replication.
Epitranscriptomic Enhancement of Influenza A Virus Gene Expression and Replication.
Cell host & microbe (2017-09-15)
David G Courtney, Edward M Kennedy, Rebekah E Dumm, Hal P Bogerd, Kevin Tsai, Nicholas S Heaton, Bryan R Cullen
ABSTRACT
Many viral RNAs are modified by methylation of the N
Sigma-Aldrich
4-Thiouridine, ≥98%
Sigma-Aldrich
Bovine Serum Albumin solution, 30% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
3-Deazaadenosine