Merck
  • Restriction endonuclease triggered bacterial apoptosis as a mechanism for long time survival.
Restriction endonuclease triggered bacterial apoptosis as a mechanism for long time survival.
Nucleic acids research (2017-09-01)
Easa Nagamalleswari, Sandhya Rao, Kommireddy Vasu, Valakunja Nagaraja
ABSTRACT
Programmed cell death (PCD) under certain conditions is one of the features of bacterial altruism. Given the bacterial diversity and varied life style, different PCD mechanisms must be operational that remain largely unexplored. We describe restriction endonuclease (REase) mediated cell death by an apoptotic pathway, beneficial for isogenic bacterial communities. Cell death is pronounced in stationary phase and when the enzyme exhibits promiscuous DNA cleavage activity. We have elucidated the molecular mechanism of REase mediated cell killing and demonstrate that released nutrients from dying cells support the growth of the remaining cells in the population. These findings illustrate a new intracellular moonlighting role for REases which are otherwise established host defence arsenals. REase induced PCD appears to be a cellular design to replenish nutrients for cells undergoing starvation stress and the phenomenon could be wide spread in bacteria, given the abundance of restriction-modification (R-M) systems in the microbial population.
Sigma-Aldrich
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)