Merck
  • Synthesis of 'reactive' and covalent polymeric multilayer coatings with durable superoleophobic and superoleophilic properties under water.
Synthesis of 'reactive' and covalent polymeric multilayer coatings with durable superoleophobic and superoleophilic properties under water.
Chemical science (2017-10-11)
Dibyangana Parbat, Uttam Manna
ABSTRACT
Bioinspired underwater super-oil-wettability (superoleophilic/superoleophobic) properties are emerging as a potential avenue for developing smart materials for addressing issues related to healthcare, environment, energy,
Sigma-Aldrich
Heptylamine, 99%