Merck
  • Photothermal detection of individual gold nanoparticles: perspectives for high-throughput screening.
Photothermal detection of individual gold nanoparticles: perspectives for high-throughput screening.
Chemphyschem : a European journal of chemical physics and physical chemistry (2008-07-31)
Florian Kulzer, Niels Laurens, Jaya Besser, Thomas Schmidt, Michel Orrit, Herman P Spaink
ABSTRACT
We use photothermal microscopy to detect and image individual gold nanoparticles that are either embedded in a polymer film or immobilized in an aqueous environment. Reducing the numerical aperture of the detection optics allows us to achieve a 200-fold-enlarged detection volume while still retaining sufficient detectivity. We characterize the capabilities of this approach for the detection of gold colloids with a diameter of 20 nm, with emphasis on practical aspects that are important for high-throughput-screening applications. The extended detection volume in combination with the stability of the photothermal signal are major advantages compared to fluorescence-based approaches, which are limited by photoblinking and photobleaching. Careful consideration is given to the trade-off between the maximum increase in local temperature that can be tolerated by a biological specimen and the minimum integration time needed to reliably determine whether a given volume contains a target species. We find that our approach has the potential to increase the detection-limited flow rate (i.e. the limit given by the detection volume divided by the minimum detection time) by two to three orders of magnitude.
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder