Merck
  • Soluble adenylyl cyclase mediates mitochondrial pathway of apoptosis and ATP metabolism in oyster Crassostrea gigas exposed to elevated CO
Soluble adenylyl cyclase mediates mitochondrial pathway of apoptosis and ATP metabolism in oyster Crassostrea gigas exposed to elevated CO
Fish & shellfish immunology (2017-05-10)
Xiudan Wang, Mengqiang Wang, Jiachao Xu, Zhihao Jia, Zhaoqun Liu, Lingling Wang, Linsheng Song
ABSTRACT
Ocean acidification (OA) has deleterious impacts on immune response and energy homeostasis status of Mollusca. In the present study, the apoptosis ratio of hemocytes and the adenosine triphosphate (ATP) allocation in gill tissues were determined after Pacific oysters Crassostrea gigas were exposed to elevated CO
Sigma-Aldrich
KH7, ≥98% (HPLC)