Merck
  • The role of interfacial lipids in stabilizing membrane protein oligomers.
The role of interfacial lipids in stabilizing membrane protein oligomers.
Nature (2017-01-13)
Kallol Gupta, Joseph A C Donlan, Jonathan T S Hopper, Povilas Uzdavinys, Michael Landreh, Weston B Struwe, David Drew, Andrew J Baldwin, Phillip J Stansfeld, Carol V Robinson
ABSTRACT
Oligomerization of membrane proteins in response to lipid binding has a critical role in many cell-signalling pathways but is often difficult to define or predict. Here we report the development of a mass spectrometry platform to determine simultaneously the presence of interfacial lipids and oligomeric stability and to uncover how lipids act as key regulators of membrane-protein association. Evaluation of oligomeric strength for a dataset of 125 α-helical oligomeric membrane proteins reveals an absence of interfacial lipids in the mass spectra of 12 membrane proteins with high oligomeric stability. For the bacterial homologue of the eukaryotic biogenic transporters (LeuT, one of the proteins with the lowest oligomeric stability), we found a precise cohort of lipids within the dimer interface. Delipidation, mutation of lipid-binding sites or expression in cardiolipin-deficient Escherichia coli abrogated dimer formation. Molecular dynamics simulation revealed that cardiolipin acts as a bidentate ligand, bridging across subunits. Subsequently, we show that for the Vibrio splendidus sugar transporter SemiSWEET, another protein with low oligomeric stability, cardiolipin shifts the equilibrium from monomer to functional dimer. We hypothesized that lipids are essential for dimerization of the Na
Sigma-Aldrich
2,2-Dimethyl-1,3-propanediol, 99%