Merck
  • Human Plasmacytoid Dendritic Cells Display and Shed B Cell Maturation Antigen upon TLR Engagement.
Human Plasmacytoid Dendritic Cells Display and Shed B Cell Maturation Antigen upon TLR Engagement.
Journal of immunology (Baltimore, Md. : 1950) (2017-03-12)
Elisabeth Schuh, Andrea Musumeci, Franziska S Thaler, Sarah Laurent, Joachim W Ellwart, Reinhard Hohlfeld, Anne Krug, Edgar Meinl
ABSTRACT
The BAFF-APRIL system is best known for its control of B cell homeostasis, and it is a target of therapeutic intervention in autoimmune diseases and lymphoma. By analyzing the expression of the three receptors of this system, B cell maturation Ag (BCMA), transmembrane activator and CAML interactor, and BAFF receptor, in sorted human immune cell subsets, we found that BCMA was transcribed in plasmacytoid dendritic cells (pDCs) in both blood and lymphoid tissue. Circulating human pDCs contained BCMA protein without displaying it on the cell surface. After engagement of TLR7/8 or TLR9, BCMA was detected also on the cell surface of pDCs. The display of BCMA on the surface of human pDCs was accompanied by release of soluble BCMA (sBCMA); inhibition of γ-secretase enhanced surface expression of BCMA and reduced the release of sBCMA by pDCs. In contrast with human pDCs, murine pDCs did not express BCMA, not even after TLR9 activation. In this study, we extend the spectrum of BCMA expression to human pDCs. sBCMA derived from pDCs might determine local availability of its high-affinity ligand APRIL, because sBCMA has been shown to function as an APRIL-specific decoy. Further, therapeutic trials targeting BCMA in patients with multiple myeloma should consider possible effects on pDCs.
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
Human TNFRSF17 ELISA Kit
Pricing and availability is not currently available.