Merck
  • Biological evaluation of 3-hydroxybenzyl alcohol, an extrolite produced by Aspergillus nidulans strain KZR-132.
Biological evaluation of 3-hydroxybenzyl alcohol, an extrolite produced by Aspergillus nidulans strain KZR-132.
Journal of applied microbiology (2017-03-18)
C Ganesh Kumar, P Mongolla, S Pombala, S Bandi, K S Babu, K V S Ramakrishna
ABSTRACT
The aim of the study was to purify and characterize a bioactive compound from Aspergillus nidulans strain KZR-132 and its biological evaluation. A bioactive extolite was purified from A. nidulans strain KZR-132, and its chemical structure was elucidated as 3-hydroxylbenzyl alcohol (3-HBA) based on This bioactive extrolite produced by A. nidulans isolated from a niche habitat was demonstrated to possess significant biotechnological and pharmacological potential since it exhibited broad-spectrum antimicrobial and antibiofilm activities which are reported for the first time. The overall study demonstrates that 3-HBA produced by A. nidulansKZR-132 is a promising bioactive metabolite and possibly can function as a pharmacologically suitable broad-spectrum antimicrobial drug candidate against various dreaded human-related bacterial and fungal pathogens.
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
3-Hydroxybenzyl alcohol, 99%