Merck
The development of novel LTA
Scientific reports (2017-03-18)
Caroline M Low, Samia Akthar, Dhiren F Patel, Stephan Löser, Chi-Tung Wong, Patricia L Jackson, J Edwin Blalock, Stephen A Hare, Clare M Lloyd, Robert J Snelgrove
ABSTRACT
The pro-inflammatory mediator leukotriene B
Sigma-Aldrich
Triacetyl resveratrol, ≥98% (HPLC)
Sigma-Aldrich
trans-Stilbene, 96%
Sigma-Aldrich
1-(2,4-Dihydroxyphenyl)-2-(4-hydroxyphenyl)ethanone, AldrichCPR