Merck
  • A novel antihuman C3d monoclonal antibody with specificity to the C3d complement split product.
A novel antihuman C3d monoclonal antibody with specificity to the C3d complement split product.
Journal of immunological methods (2017-02-09)
Karina Juhl Rasmussen, Mikkel-Ole Skjoedt, Lars Vitved, Karsten Skjoedt, Yaseelan Palarasah
ABSTRACT
The complement component C3 and the cleavage products of C3b/iC3b, C3c and C3d are used as biomarkers in clinical diagnostics. Currently, no specific antibodies are able to differentiate C3d from other fragments, although such a distinction could be very valuable considering that they may reflect different pathophysiological mechanisms. We have developed a rat antihuman C3d monoclonal antibody with specificity to the end sequence of the N-terminal region of C3d. The antibody can therefore only bind to C3d when it manifests itself as the final end product of cleaved C3. We believe that this specificity is it first of its kind, and predicts that it can be used as a detection tool in several immunological methods with great value in diagnostics.
Sigma-Aldrich
Snake venom from Naja naja kaouthia