Merck
  • Istamycin aminoglycosides profiling and their characterization in Streptomyces tenjimariensis ATCC 31603 culture using high-performance liquid chromatography with tandem mass spectrometry.
Istamycin aminoglycosides profiling and their characterization in Streptomyces tenjimariensis ATCC 31603 culture using high-performance liquid chromatography with tandem mass spectrometry.
Journal of separation science (2016-10-26)
Nguyen Huu Hoang, Nguyen Lan Huong, Byul Kim, Jae Kyung Sohng, Yeo Joon Yoon, Je Won Park
ABSTRACT
A high-performance liquid chromatography with electrospray ionization ion trap tandem mass spectrometry method was developed and validated for the robust profiling and characterization of biosynthetic congeners in the 2-deoxy-aminocyclitol istamycin pathway, from the fermentation broth of Streptomyces tenjimariensis ATCC 31603. Gradient elution on an Acquity CSH C
Sigma-Aldrich
Pentafluoropropionic acid, 97%