- New cobalt(II) and nickel(II) complexes of benzyl carbazate Schiff bases: Syntheses, crystal structures, in vitro DNA and HSA binding studies.
New cobalt(II) and nickel(II) complexes of benzyl carbazate Schiff bases: Syntheses, crystal structures, in vitro DNA and HSA binding studies.
Journal of photochemistry and photobiology. B, Biology (2016-11-07)
Palanivelu Nithya, Sannasi Helena, Jim Simpson, Malaichamy Ilanchelian, Aathi Muthusankar, Subbiah Govindarajan
PMID27816644
ABSTRACT
In the present study, new Schiff base complexes with the composition [M(NCS)
MATERIALS