Merck
Stable Low-Bandgap Pb-Sn Binary Perovskites for Tandem Solar Cells.
Advanced materials (Deerfield Beach, Fla.) (2016-10-21)
Zhibin Yang, Adharsh Rajagopal, Chu-Chen Chueh, Sae Byeok Jo, Bo Liu, Ting Zhao, Alex K-Y Jen
ABSTRACT
A low-bandgap (1.33 eV) Sn-based MA
Sigma-Aldrich
Tin(IV) iodide, anhydrous, powder, 99.999% trace metals basis