Merck
  • Diving into the Finestructure of Macroporous Polymer Foams Synthesized via Emulsion Templating: A Phase Diagram Study.
Diving into the Finestructure of Macroporous Polymer Foams Synthesized via Emulsion Templating: A Phase Diagram Study.
Langmuir : the ACS journal of surfaces and colloids (2017-01-07)
Aggeliki Quell, Thomas Sottmann, Cosima Stubenrauch
ABSTRACT
During our studies on emulsion-templated monodisperse polymer foams we found significant differences in the finestructure if the locus of initiation is changed. This motivated us to study the phase behavior of the liquid template. Our studies indicate that the template consists of droplets of three different length scales: The water droplets generated via microfluidics (∼70 μm) are surrounded by a continuous phase in which a w/o emulsion (≤100 nm) coexists with a w/o microemulsion (∼5 nm). We speculate that the w/o-emulsion droplets act as seeds for the porous finestructure observed in AIBN-initiated polymer foams. We have experimental evidence that the w/o emulsion inverts to an o/w emulsion with progressing polymerization. This explains the granular texture observed in KPS-initiated polymer foams. The control of the finestructure is important in the preparation of tailor-made polymer foams because it directly impacts the material's density and thus, in turn, its mechanical stability.
Sigma-Aldrich
Octaethylene glycol monodecyl ether, BioXtra, ≥98.0% (TLC)