Merck
  • Pyrogallol abates VSMC migration via modulation of Caveolin-1, matrix metalloproteinase and intima hyperplasia in carotid ligation mouse.
Pyrogallol abates VSMC migration via modulation of Caveolin-1, matrix metalloproteinase and intima hyperplasia in carotid ligation mouse.
Environmental toxicology and pharmacology (2016-10-22)
Yu-Dong Ma, Varadharajan Thiyagarajan, May-Jywan Tsai, Sheng-I Lue, Yi-Chen Chia, Song-Kun Shyue, Ching-Feng Weng
ABSTRACT
Migration of vascular smooth muscle cells (VSMCs) contributes to intimal hyperplasia and other vascular diseases. Caveolin-1 (Cav-1) has been recognized as a proliferative inhibitor of VSMCs and is likely to be an important regulator of VSMC migration. The underlying mechanism of pyrogallol on the VSMC migration is not fully understood. This study attempted to dissect the role of Cav-1 and matrix metalloproteinase (MMP) in VSMC migration and to investigate the effect of pyrogallol on VSMC mobility during carotid artery ligation mice. The mRNA expression of MMP-3 and MMP-13 was down-regulated in cultured VSMC prepared from Cav-1-deficient (Cav-1 KO) mice whereas MMP-14 expression was up-regulated. Pyrogallol effectively inhibited the migration of Cav-1 KO VSMC by promoting the expression of tissue inhibitors of metalloproteinase (TIMP)-2. Pyrogallol also inhibited the migration of Cav-1 wild type (WT) VSMC, however, by increasing TIMP-1 expression and repressing MMP-2 activity. In a parallel in vivo study, intra-peritoneal (ip) of pyrogallol to carotid artery ligated mice significantly suppressed intima formation in mice carotid artery. Furthermore, the proMMP-9 activity in pyrogallol-treated mice serum significantly increased from Day 0 to Day 2 and decreased from Day 2 to Day 7 in a time-dependent manner. In addition, WT mice treated with pyrogallol had significantly reduced neointima formation, whereas no differences were observed in Cav-1 knock out (KO) mice. These results suggest that pyrogallol not only inhibited VSMC migration but also effectively diminishes the severity of neointima hyperplasia, implying that pyrogallol possesses potential anti-atherogenic effects for the treatment of vascular diseases.
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A