Merck
  • Ginger extract protects rat's kidneys against oxidative damage after chronic ethanol administration.
Ginger extract protects rat's kidneys against oxidative damage after chronic ethanol administration.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-10-07)
Aireza Shirpoor, Farzaneh Rezaei, Amin Abdollahzade Fard, Ali Taghizadeh Afshari, Farzaneh Hosseini Gharalari, Yousef Rasmi
ABSTRACT
Chronic alcohol ingestion is associated with pronounced detrimental effects on the renal system. In the current study, the protective effect of ginger extract on ethanol-induced damage was evaluated through determining 8-OHdG, cystatin C, glomerular filtration rate, and pathological changes such as cell proliferation and fibrosis in rats' kidneys. Male wistar rats were randomly divided into three groups and were treated as follows: (1) control, (2) ethanol and (3) ginger extract treated ethanolic (GETE) groups. After a six weeks period of treatment, the results revealed proliferation of glomerular and tubular cells, fibrosis in glomerular and peritubular and a significant rise in the level of 8-OHdG, cystatin C, plasma urea and creatinine. Moreover, compared to the control group, the ethanol group showed a significant decrease in the urine creatinine and creatinine clearance. In addition, significant amelioration of changes in the structure of kidneys, along with restoration of the biochemical alterations were found in the ginger extract treated ethanolic group, compared to the ethanol group. These findings indicate that ethanol induces kidneys abnormality by oxidative DNA damage and oxidative stress, and that these effects can be alleviated using ginger as an antioxidant and anti-inflammatory agent.
Sigma-Aldrich
8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)