Merck
  • Therapeutic drug monitoring of eculizumab: Rationale for an individualized dosing schedule.
Therapeutic drug monitoring of eculizumab: Rationale for an individualized dosing schedule.
mAbs (2015-09-05)
Philippe Gatault, Guillaume Brachet, David Ternant, Danielle Degenne, Guillaume Récipon, Christelle Barbet, Emmanuel Gyan, Valérie Gouilleux-Gruart, Cécile Bordes, Alexandra Farrell, Jean Michel Halimi, Hervé Watier
ABSTRACT
The annual cost of eculizumab maintenance therapy in paroxysmal nocturnal hemoglobinuria (PNH) and atypical hemolytic-uremic syndrome (aHUS) exceeds $300,000 per patient. A better understanding of eculizumab pharmacokinetics and subsequent individual dose adjustment could reduce this cost. We measured the trough eculizumab concentration in 9 patients with maintenance therapy (aHUS, n = 7; PNH, n = 2) and determined: 1) the intra- and inter-individual variability; 2) the influence of weight on eculizumab pharmacokinetics; and 3) the rate of elimination of eculizumab following discontinuation. A one-compartment model was developed to describe the pharmacokinetics of eculizumab and predicted complement activity by body weight. Trough eculizumab concentrations were >50 µg/mL in 9/9, >100 µg/mL in 8/9, and >300 µg/mL in 5/9 of patients. Intra-individual variability was low but eculizumab concentrations, closely correlated with patient weight (R(2) = 0.66, p = 0.034), varied broadly (55 ± 12 to 733 ± 164 µg/mL). Pharmacokinetic modeling showed that the elimination half-life varied greatly, with an increase from 7.8 d in a patient weighing 100 kg to 19.5 d in a 40 kg patient. We predicted that infusions of 1200 mg could be spaced every 4 or 6 weeks in patients weighing <90 and <70 kg, respectively. In this pilot study, the current recommended use of a fixed eculizumab dose for maintenance therapy is associated with excessively high trough concentrations in many patients. Further prospective larger studies are now required to support an individualized schedule adjusted for patient weight and based on the observed trough serum eculizumab concentration.
Sigma-Aldrich
Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution