Merck
  • Apurinic/Apyrimidinic Endonuclease 1/Redox Factor-1 (Ape1/Ref-1) Modulates Antigen Presenting Cell-mediated T Helper Cell Type 1 Responses.
Apurinic/Apyrimidinic Endonuclease 1/Redox Factor-1 (Ape1/Ref-1) Modulates Antigen Presenting Cell-mediated T Helper Cell Type 1 Responses.
The Journal of biological chemistry (2016-09-18)
Nasrin Akhter, Yuji Takeda, Hidetoshi Nara, Akemi Araki, Naoto Ishii, Naoki Asao, Hironobu Asao
ABSTRACT
Apurinic/apyrimidinic endonuclease 1/redox factor-1 (Ape1/Ref-1) is a multifunctional protein possessing DNA repair, redox control, and transcriptional regulatory activities. Although Ape1/Ref-1 plays multiple roles in the immune system, its functions in helper T (Th) cell activation and differentiation are largely unknown. In this study, the function of Ape1/Ref-1 in Th cell activation was analyzed using an Ape1/Ref-1 redox-specific inhibitor, E3330. When splenocytes from OT-II mice, which are ovalbumin (OVA)-specific T-cell receptor transgenic mice, were activated with OVA in the presence of E3330, the induction of IFN-γ-producing OT-II T cells was significantly increased. In contrast, E3330 did not enhance IFN-γ production from plate-bound anti-CD3 antibody-stimulated CD4
Sigma-Aldrich
MISSION® esiRNA, targeting human APEX1