Merck
  • Chiral recognition of penicillamine enantiomers using hemoglobin and gold nanoparticles functionalized graphite-like carbon nitride nanosheets via electrochemiluminescence.
Chiral recognition of penicillamine enantiomers using hemoglobin and gold nanoparticles functionalized graphite-like carbon nitride nanosheets via electrochemiluminescence.
Colloids and surfaces. B, Biointerfaces (2016-11-05)
Xia Lin, Shu Zhu, Qinghong Wang, Qiao Xia, Peiyao Ran, Yingzi Fu
ABSTRACT
A new stable and stereo-selective electrochemiluminescence (ECL) interface has been designed for specific recognition of penicillamine (Pen) enantiomers by using hemoglobin (Hb) and gold nanoparticles functionalized graphite-like carbon nitride nanosheets composite (Au-g-C
Sigma-Aldrich
L-Penicillamine, 99%