Merck
  • New ways to synthesize lead sulfide nanosheets-substituted alkanes direct the growth of 2D nanostructures.
New ways to synthesize lead sulfide nanosheets-substituted alkanes direct the growth of 2D nanostructures.
Nanotechnology (2016-07-28)
Thomas Bielewicz, Eugen Klein, Christian Klinke
ABSTRACT
Two-dimensional colloidal nanosheets represent very attractive optoelectronic materials. They combine good lateral conductivity with solution-processability and geometry-tunable electronic properties. In the case of PbS nanosheets, so far synthesis has been driven by the addition of chloroalkanes as coligands. Here, we demonstrate how to synthesize two-dimensional lead sulfide nanostructures using other halogen alkanes and primary amines. Further, we show that at a reaction temperature of 170 °C a coligand is not even necessary and the only ligand, oleic acid, controls the anisotropic growth of the two-dimensional structures. Also, using thiourea as a sulfide source, nanosheets with lateral dimensions of over 10 μm are possible.
Sigma-Aldrich
1-Chloroheptane, 99%
Sigma-Aldrich
1-Iododecane, 98%