Merck
  • Asymmetric counteranion-directed Lewis acid organocatalysis for the scalable cyanosilylation of aldehydes.
Asymmetric counteranion-directed Lewis acid organocatalysis for the scalable cyanosilylation of aldehydes.
Nature communications (2016-08-18)
Zhipeng Zhang, Han Yong Bae, Joyram Guin, Constantinos Rabalakos, Manuel van Gemmeren, Markus Leutzsch, Martin Klussmann, Benjamin List
ABSTRACT
Due to the high versatility of chiral cyanohydrins, the catalytic asymmetric cyanation reaction of carbonyl compounds has attracted widespread interest. However, efficient protocols that function at a preparative scale with low catalyst loading are still rare. Here, asymmetric counteranion-directed Lewis acid organocatalysis proves to be remarkably successful in addressing this problem and enabled a molar-scale cyanosilylation in quantitative yield and with excellent enantioselectivity. Also, the catalyst loading could be lowered to a part-per-million level (50 ppm: 0.005 mol%). A readily accessible chiral disulfonimide was used, which in combination with trimethylsilyl cyanide, turned into the active silylium Lewis acid organocatalyst. The nature of a peculiar phenomenon referred to as a "dormant period", which is mainly induced by water, was systematically investigated by means of in situ Fourier transform infrared analysis.
Sigma-Aldrich
2-Naphthaldehyde, 98%