Merck
  • Glucocorticoid antagonism limits adiposity rebound and glucose intolerance in young male rats following the cessation of daily exercise and caloric restriction.
Glucocorticoid antagonism limits adiposity rebound and glucose intolerance in young male rats following the cessation of daily exercise and caloric restriction.
American journal of physiology. Endocrinology and metabolism (2016-05-05)
Trevor Teich, Emily C Dunford, Deanna P Porras, Jacklyn A Pivovarov, Jacqueline L Beaudry, Hazel Hunt, Joseph K Belanoff, Michael C Riddell
ABSTRACT
Severe caloric restriction (CR), in a setting of regular physical exercise, may be a stress that sets the stage for adiposity rebound and insulin resistance when the food restriction and exercise stop. In this study, we examined the effect of mifepristone, a glucocorticoid (GC) receptor antagonist, on limiting adipose tissue mass gain and preserving whole body insulin sensitivity following the cessation of daily running and CR. We calorically restricted male Sprague-Dawley rats and provided access to voluntary running wheels for 3 wk followed by locking of the wheels and reintroduction to ad libitum feeding with or without mifepristone (80 mg·kg(-1)·day(-1)) for 1 wk. Cessation of daily running and CR increased HOMA-IR and visceral adipose mass as well as glucose and insulin area under the curve during an oral glucose tolerance test vs. pre-wheel lock exercised rats and sedentary rats (all P < 0.05). Insulin sensitivity and glucose tolerance were preserved and adipose tissue mass gain was attenuated by daily mifepristone treatment during the post-wheel lock period. These findings suggest that following regular exercise and CR there are GC-induced mechanisms that promote adipose tissue mass gain and impaired metabolic control in healthy organisms and that this phenomenon can be inhibited by the GC receptor antagonist mifepristone.
Sigma-Aldrich
Rat CRP/C-Reactive Protein ELISA Kit, for serum, plasma and cell culture supernatant