Merck
  • Alkylation of active methylene compounds with alcohols catalyzed by an iridium complex.
Alkylation of active methylene compounds with alcohols catalyzed by an iridium complex.
Chemical communications (Cambridge, England) (2007-07-05)
Masao Morita, Yasushi Obora, Yasutaka Ishii
ABSTRACT
Base-free catalytic alpha-alkylation of active methylene compounds with primary alcohols was successfully achieved using an [IrCl(cod)](2) complex in the presence of PPh(3) to afford the corresponding saturated alpha-alkylated products in good yields.
Sigma-Aldrich
2-(Benzyloxy)ethanol, 98%