Merck
  • Synthesis and pharmacological evaluation of new 1,2-dithiolane based antioxidants.
Synthesis and pharmacological evaluation of new 1,2-dithiolane based antioxidants.
European journal of medicinal chemistry (2003-02-21)
Claude Guillonneau, Yves Charton, Yves-Michel Ginot, Marie-Victoire Fouquier-d'Hérouël, Marc Bertrand, Brian Lockhart, Pierre Lestage, Solo Goldstein
ABSTRACT
Molecules containing a dithiolane moiety are widely investigated due to their antioxidant properties. The archetypal representative of this class of compounds is lipoic acid and indeed the lipoic acid-dihydrolipoic acid couple is part of the antioxidant defence system of the cell. In the course of a program aiming to find improved antioxidants effective in vivo, we designed, synthesised and pharmacologically investigated new lipoic acid analogs. The salient feature of these structures is the connection, via a thioamide or a thiocarbamate, of a 1,2-dithiolane moiety bearing a carbon chain and a N-alkyl-substituted morpholine ring. It was expected that the antioxidant and chelating properties of these functional groups combined with the basicity of the morpholine ring will impact on the antioxidant as well as on the partition and solubility characteristics of the compounds. Indeed in vitro and in vivo pharmacological investigation showed that these new molecules and especially those containing a thiocarbamate linker possess superior antioxidant properties compared with alpha-lipoic acid and to the amide or carbamate linker analogs. In particular, some of these compounds efficiently cross the blood brain barrier (BBB) thus providing efficient protection from lethality in a situation of induced oxidative stress. Moreover the absence of the 1,2-dithiolane moiety does not completely abolish antioxidant effects thus demonstrating that these compounds are distinct new chemical entities and not merely lipoic acid prodrugs. The chemical and pharmacological features of these new antioxidants are presented and discussed in the following paper.
Sigma-Aldrich
2-(4-Morpholino)ethyl isothiocyanate, 96%
Pricing and availability is not currently available.