Merck
  • Role for the thromboxane A2 receptor β-isoform in the pathogenesis of intrauterine growth restriction.
Role for the thromboxane A2 receptor β-isoform in the pathogenesis of intrauterine growth restriction.
Scientific reports (2016-07-02)
Katie L Powell, Veronica Stevens, Dannielle H Upton, Sharon A McCracken, Ann M Simpson, Yan Cheng, Vitomir Tasevski, Jonathan M Morris, Anthony W Ashton
ABSTRACT
Intrauterine growth restriction (IUGR) is a pathology of pregnancy that results in failure of the fetus to reach its genetically determined growth potential. In developed nations the most common cause of IUGR is impaired placentation resulting from poor trophoblast function, which reduces blood flow to the fetoplacental unit, promotes hypoxia and enhances production of bioactive lipids (TXA2 and isoprostanes) which act through the thromboxane receptor (TP). TP activation has been implicated as a pathogenic factor in pregnancy complications, including IUGR; however, the role of TP isoforms during pregnancy is poorly defined. We have determined that expression of the human-specific isoform of TP (TPβ) is increased in placentae from IUGR pregnancies, compared to healthy pregnancies. Overexpression of TPα enhanced trophoblast proliferation and syncytialisation. Conversely, TPβ attenuated these functions and inhibited migration. Expression of the TPβ transgene in mice resulted in growth restricted pups and placentae with poor syncytialisation and diminished growth characteristics. Together our data indicate that expression of TPα mediates normal placentation; however, TPβ impairs placentation, and promotes the development of IUGR, and represents an underappreciated pathogenic factor in humans.
Sigma-Aldrich
L-Mimosine from Koa hoale seeds, ≥98%
Pricing and availability is not currently available.