Merck
Opioid metabolism.
Mayo Clinic proceedings (2009-07-02)
Howard S Smith
ABSTRACT
Clinicians understand that individual patients differ in their response to specific opioid analgesics and that patients may require trials of several opioids before finding an agent that provides effective analgesia with acceptable tolerability. Reasons for this variability include factors that are not clearly understood, such as allelic variants that dictate the complement of opioid receptors and subtle differences in the receptor-binding profiles of opioids. However, altered opioid metabolism may also influence response in terms of efficacy and tolerability, and several factors contributing to this metabolic variability have been identified. For example, the risk of drug interactions with an opioid is determined largely by which enzyme systems metabolize the opioid. The rate and pathways of opioid metabolism may also be influenced by genetic factors, race, and medical conditions (most notably liver or kidney disease). This review describes the basics of opioid metabolism as well as the factors influencing it and provides recommendations for addressing metabolic issues that may compromise effective pain management. Articles cited in this review were identified via a search of MEDLINE, EMBASE, and PubMed. Articles selected for inclusion discussed general physiologic aspects of opioid metabolism, metabolic characteristics of specific opioids, patient-specific factors influencing drug metabolism, drug interactions, and adverse events.