Merck
  • Genome sequence of Xanthomonas fuscans subsp. fuscans strain 4834-R reveals that flagellar motility is not a general feature of xanthomonads.
Genome sequence of Xanthomonas fuscans subsp. fuscans strain 4834-R reveals that flagellar motility is not a general feature of xanthomonads.
BMC genomics (2013-11-08)
Armelle Darrasse, Sébastien Carrère, Valérie Barbe, Tristan Boureau, Mario L Arrieta-Ortiz, Sophie Bonneau, Martial Briand, Chrystelle Brin, Stéphane Cociancich, Karine Durand, Stéphanie Fouteau, Lionel Gagnevin, Fabien Guérin, Endrick Guy, Arnaud Indiana, Ralf Koebnik, Emmanuelle Lauber, Alejandra Munoz, Laurent D Noël, Isabelle Pieretti, Stéphane Poussier, Olivier Pruvost, Isabelle Robène-Soustrade, Philippe Rott, Monique Royer, Laurana Serres-Giardi, Boris Szurek, Marie-Anne van Sluys, Valérie Verdier, Christian Vernière, Matthieu Arlat, Charles Manceau, Marie-Agnès Jacques
ABSTRACT
Xanthomonads are plant-associated bacteria responsible for diseases on economically important crops. Xanthomonas fuscans subsp. fuscans (Xff) is one of the causal agents of common bacterial blight of bean. In this study, the complete genome sequence of strain Xff 4834-R was determined and compared to other Xanthomonas genome sequences. Comparative genomics analyses revealed core characteristics shared between Xff 4834-R and other xanthomonads including chemotaxis elements, two-component systems, TonB-dependent transporters, secretion systems (from T1SS to T6SS) and multiple effectors. For instance a repertoire of 29 Type 3 Effectors (T3Es) with two Transcription Activator-Like Effectors was predicted. Mobile elements were associated with major modifications in the genome structure and gene content in comparison to other Xanthomonas genomes. Notably, a deletion of 33 kbp affects flagellum biosynthesis in Xff 4834-R. The presence of a complete flagellar cluster was assessed in a collection of more than 300 strains representing different species and pathovars of Xanthomonas. Five percent of the tested strains presented a deletion in the flagellar cluster and were non-motile. Moreover, half of the Xff strains isolated from the same epidemic than 4834-R was non-motile and this ratio was conserved in the strains colonizing the next bean seed generations. This work describes the first genome of a Xanthomonas strain pathogenic on bean and reports the existence of non-motile xanthomonads belonging to different species and pathovars. Isolation of such Xff variants from a natural epidemic may suggest that flagellar motility is not a key function for in planta fitness.
Sigma-Aldrich
Gly-Pro-Glu, ≥98% (HPLC)