Merck
  • Effects of Lactation Stage and Individual Performance on Milk cis-9, trans-11 Conjugated Linoleic Acids Content in Dairy Cows.
Effects of Lactation Stage and Individual Performance on Milk cis-9, trans-11 Conjugated Linoleic Acids Content in Dairy Cows.
Asian-Australasian journal of animal sciences (2013-02-01)
T Wang, J J Oh, J N Lim, J E Hong, J H Kim, J H Kim, H S Kang, Y J Choi, H G Lee
ABSTRACT
The goal of this study was to evaluate the effects of lactation stage and individual performance on milk cis-9, trans-11 conjugated linoleic acid (CLA) content in dairy cows. In experiment 1, the milk cis-9, trans-11 CLA content from dairy cows in early (0.33±0.014%), middle (0.37±0.010%), and late stages (0.44±0.020%) showed significant differences (p<0.05); and the individual contents of the major fatty acids, especially cis-9, trans-11 CLA in cows of the same lactation were also variable. In the second experiment design as a validation test, our results once again showed that the individual contents of cis-9, trans-11 CLA were various, and a difference of about 2-fold (0.55% vs 0.95%) was observed, although the animals were offered same diet. These data demonstrated that lactation stage and individual performance have considerable effects on milk cis-9, trans-11 CLA contents.
Sigma-Aldrich
Methyl trans-vaccenate, ≥99% (capillary GC), liquid
Pricing and availability is not currently available.