Merck
Melanopsin mediates retrograde visual signaling in the retina.
PloS one (2012-08-11)
Dao-Qi Zhang, Michael A Belenky, Patricia J Sollars, Gary E Pickard, Douglas G McMahon
ABSTRACT
The canonical flow of visual signals proceeds from outer to inner retina (photoreceptors → bipolar cells → ganglion cells). However, melanopsin-expressing ganglion cells are photosensitive and functional sustained light signaling to retinal dopaminergic interneurons persists in the absence of rods and cones. Here we show that the sustained-type light response of retinal dopamine neurons requires melanopsin and that the response is mediated by AMPA-type glutamate receptors, defining a retrograde retinal visual signaling pathway that fully reverses the usual flow of light signals in retinal circuits.
Sigma-Aldrich
L-(+)-2-Amino-4-phosphonobutyric acid, optical purity optical purity: ≥95% (HPLC, Marfey′s reagent)