Merck
  • Gene transfer of mutant mouse cholinesterase provides high lifetime expression and reduced cocaine responses with no evident toxicity.
Gene transfer of mutant mouse cholinesterase provides high lifetime expression and reduced cocaine responses with no evident toxicity.
PloS one (2013-07-11)
Liyi Geng, Yang Gao, Xiabin Chen, Shurong Hou, Chang-Guo Zhan, Zoran Radic, Robin J Parks, Stephen J Russell, Linh Pham, Stephen Brimijoin
ABSTRACT
Gene transfer of a human cocaine hydrolase (hCocH) derived from butyrylcholinesterase (BChE) by 5 mutations (A199S/F227A/S287G/A328W/Y332G) has shown promise in animal studies for treatment of cocaine addiction. To predict the physiological fate and immunogenicity of this enzyme in humans, a comparable enzyme was created and tested in a conspecific host. Thus, similar mutations (A199S/S227A/S287G/A328W/Y332G) were introduced into mouse BChE to obtain a mouse CocH (mCocH). The cDNA was incorporated into viral vectors based on: a) serotype-5 helper-dependent adenovirus (hdAD) with ApoE promoter, and b) serotype-8 adeno-associated virus with CMV promoter (AAV-CMV) or multiple promoter and enhancer elements (AAV-VIP). Experiments on substrate kinetics of purified mCocH expressed in HEK293T cells showed 30-fold higher activity (U/mg) with (3)H-cocaine and 25% lower activity with butyrylthiocholine, compared with wild type BChE. In mice given modest doses of AAV-CMV-mCocH vector (0.7 or 3 × 10(11) particles) plasma hydrolase activity rose 10-fold above control for over one year with no observed immune response. Under the same conditions, transduction of the human counterpart continued less than 2 months and antibodies to hCocH were readily detected. The advanced AAV-VIP-mCocH vector generated a dose-dependent rise in plasma cocaine hydrolase activity from 20-fold (10(10) particles) to 20,000 fold (10(13) particles), while the hdAD vector (1.7 × 10(12) particles) yielded a 300,000-fold increase. Neither vector caused adverse reactions such as motor weakness, elevated liver enzymes, or disturbance in spontaneous activity. Furthermore, treatment with high dose hdAD-ApoE-mCocH vector (1.7 × 10(12) particles) prevented locomotor abnormalities, other behavioral signs, and release of hepatic alanine amino transferase after a cocaine dose fatal to most control mice (120 mg/kg). This outcome suggests that viral gene transfer can yield clinically effective cocaine hydrolase expression for lengthy periods without immune reactions or cholinergic dysfunction, while blocking toxicity from drug overdose.