Merck
  • GITR subverts Foxp3(+) Tregs to boost Th9 immunity through regulation of histone acetylation.
GITR subverts Foxp3(+) Tregs to boost Th9 immunity through regulation of histone acetylation.
Nature communications (2015-09-15)
Xiang Xiao, Xiaomin Shi, Yihui Fan, Xiaolong Zhang, Minhao Wu, Peixiang Lan, Laurie Minze, Yang-Xin Fu, Rafik M Ghobrial, Wentao Liu, Xian Chang Li
ABSTRACT
Glucocorticoid-induced TNFR-related protein (GITR) is a costimulatory molecule with diverse effects on effector T cells and regulatory T cells (Tregs), but the underlying mechanism remains poorly defined. Here we demonstrate that GITR ligation subverts the induction of Foxp3(+) Tregs and directs the activated CD4(+) T cells to Th9 cells. Such GITR-mediated iTreg to Th9 induction enhances anti-tumour immunity in vivo. Mechanistically, GITR upregulates the NF-κB family member p50, which recruits histone deacetylases to the Foxp3 locus to produce a 'closed' chromatin structure. Furthermore, GITR ligation also activates STAT6, and STAT6 renders Il9 locus accessible via recruitment of histone acetyltransferase p300, and together with inhibition of Foxp3, GITR induces strong Th9 responses. Thus, Th9 cells and iTregs are developmentally linked and GITR can subvert tolerogenic conditions to boost Th9 immunity.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Protein G Agarose/Salmon Sperm DNA, 2.5 mL, for use in chromatin immunoprecipitations (ChIP assays)
Sigma-Aldrich
Anti-dimethyl-Histone H3 (diMe-Lys9) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution