Merck
  • Synthesis and structure-activity relationships of some 6 beta-acrylamido penicillins.
Synthesis and structure-activity relationships of some 6 beta-acrylamido penicillins.
The Journal of antibiotics (1993-02-01)
R K Anderson, P C Chapman, S C Cosham, J S Davies, T J Grinter, M A Harris, D J Merrikin, C A Mitchell, R J Ponsford, C F Smith
ABSTRACT
Syntheses are described for penicillins (4b approximately 4i, 5a and 5b) which possess a 6 beta-(2-heteroaryl-3-substituted)-propenamido side-chain of fixed geometry. In vitro results for these compounds against a range of Gram-positive and Gram-negative bacteria showed in most cases good stability against both penicillinase and TEM-1 beta-lactamase; analogues (4b approximately 4i) bearing a 2-(2-aminothiazol-4-yl) unit showed the best intrinsic activity, the cyclohexyl compound (4b) being the most promising. The 1-acetoxyethyl ester (6) of 4b was also prepared; in experimental animal studies the in vivo properties of this compound compared favourably with cefuroxime axetil and are reported together with selected in vivo data for the other compounds.
Sigma-Aldrich
Tetrahydro-4H-pyran-4-one, 99%
Pricing and availability is not currently available.